Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007725.2 | NP_031751.1 | 1, 2 | 142 | ||
GenBank mRNA | AK051845.1 | - | 1, 2 | 143 | ||
AK189304.1 | - | 1, 2 | 70 | |||
BC008181.1 | - | 1, 2 | 60 | 3589470 | ||
BC009144.1 | - | 1, 2 | 97 | 3594856 | ||
BC018482.1 | - | 1, 2 | 112 | 3589158 |