Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136056.2 | NP_001129528.1 | 7 | 933 | ||
NM_016673.2 | NP_057882.2 | 7 | 912 | |||
GenBank mRNA | AB154246.1 | - | 5 | 708 | ||
AF068615.1 | - | 7 | 863 | |||
AK137026.1 | - | 7 | 822 | |||
AK194669.1 | - | 1 | 72 | |||
BC046974.1 | - | 7 | 902 | 6389656 | ||
BC050928.1 | - | 7 | 930 | 6415187 |