Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col10a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009925.4 | NP_034055.1 | 3 | 1910 | ||
GenBank mRNA | X63013.1 | - | 1 | 680 |