Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007729.2 | NP_031755.2 | 6, 7 | 1258 | ||
GenBank mRNA | AK140136.1 | - | 6, 7 | 1257 | ||
AK157734.1 | - | 6, 7 | 1102 | |||
BC052161.1 | - | 6, 7 | 1208 | 6393164 | ||
D38162.1 | - | 6, 7 | 1072 |