Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007737.2 | NP_031763.2 | 54 | 4826 | ||
GenBank mRNA | AK084285.1 | - | 16 | 1643 | ||
AK084538.1 | - | 13 | 1610 | |||
AK084803.1 | - | 54 | 4465 | |||
AK132413.1 | - | 54 | 4608 | |||
AK139130.1 | - | 54 | 4401 | |||
AK147220.1 | - | 54 | 4608 | |||
AK147328.1 | - | 54 | 4827 | |||
AK151929.1 | - | 54 | 4610 | |||
AK160008.1 | - | 54 | 4400 | |||
BC043696.1 | - | 28 | 2641 | |||
BC055077.1 | - | 54 | 4577 | 30059991 | ||
L02918.1 | - | 54 | 4458 |