Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007748.3 | NP_031774.1 | 1, 2 | 115 | ||
GenBank mRNA | AK002402.1 | - | 1, 2 | 117 | ||
AK019318.1 | - | 1, 2 | 120 | |||
AK166496.1 | - | 1, 2 | 117 | |||
BC003807.1 | - | 1, 2 | 115 | 3487598 | ||
BC052816.1 | - | 1, 2 | 121 | 6439987 | ||
L06465.1 | - | 1, 2 | 114 | |||
U08440.1 | - | 1, 2 | 104 |