Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009943.2 | NP_034073.2 | 2 | 212 | ||
GenBank mRNA | AK002506.1 | - | 2 | 163 | ||
BC028514.1 | - | 2 | 183 | 1447375 | ||
BF472154.1 | - | 2 | 95 | |||
BU842968.1 | - | 2 | 212 | |||
U08439.1 | - | 2 | 198 |