Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009945.3 | NP_034075.2 | 4 | 406 | ||
GenBank mRNA | AF037371.1 | - | 4 | 280 | ||
AK007875.1 | - | 4 | 283 | |||
AK011269.1 | - | 4 | 266 | |||
AK019210.1 | - | 4 | 261 | |||
AK153173.1 | - | 4 | 283 | |||
BC010979.1 | - | 4 | 251 | 4217409 | ||
CN670202.1 | - | 4 | 269 | |||
CN838002.1 | - | 4 | 409 | |||
X58486.1 | - | 4 | 238 |