Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276248.1 | NP_001263177.1 | 5 | 1222 | ||
NM_001276250.1 | NP_001263179.1 | 5 | 1222 | |||
NM_007752.3 | NP_031778.2 | 5 | 1222 | |||
GenBank mRNA | AK014835.1 | - | 5 | 1208 | ||
AK043248.1 | - | 5 | 1167 | |||
AK160035.1 | - | 1 | 148 | |||
BC062957.1 | - | 5 | 1222 | 30547760 | ||
DQ336396.1 | - | 5 | 1220 | |||
U49430.1 | - | 5 | 1097 |