Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cplx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007756.3 | NP_031782.3 | 3 | 349 | ||
GenBank mRNA | AK134820.1 | - | 3 | 292 | ||
AK158166.1 | - | 3 | 291 | |||
AK159026.1 | - | 3 | 308 | |||
BC014803.1 | - | 3 | 254 | 4527117 | ||
D38614.1 | - | 3 | 281 |