Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009948.2 | NP_034078.2 | 18 | 2299 | ||
GenBank mRNA | AB010826.1 | - | 17 | 2234 | ||
AF017174.2 | - | 18 | 2299 | |||
AK029141.1 | - | 18 | 2275 | |||
AK146851.1 | - | 18 | 2301 | |||
BC018270.1 | - | 18 | 2266 | 4920625 | ||
CJ238532.1 | - | 2 | 85 |