Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007760.3 | NP_031786.2 | 5 | 979 | ||
GenBank mRNA | AK041431.1 | - | 5 | 754 | ||
AK132179.1 | - | 6 | 806 | |||
AK170740.1 | - | 5 | 765 | |||
BC006668.1 | - | 5 | 726 | 3499169 | ||
X85983.1 | - | 5 | 697 |