Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110851.1 | NP_001104321.1 | 3, 4 | 473 | ||
NM_001110853.2 | NP_001104323.1 | 2, 3 | 252 | |||
NM_001110856.2 | NP_001104326.1 | 3, 4 | 361 | |||
NM_001110859.2 | NP_001104329.1 | 3, 4 | 361 | |||
NM_001271503.1 | NP_001258432.1 | 3, 4 | 361 | |||
NM_001271504.1 | NP_001258433.1 | 3, 4 | 361 | |||
NM_001271506.1 | NP_001258435.1 | 3, 4 | 361 | |||
GenBank mRNA | AK015641.1 | - | 2, 3 | 253 | ||
AK016156.1 | - | 2, 3 | 239 | |||
AK076797.1 | - | 2, 3 | 175 | |||
AK161366.1 | - | 3, 4 | 407 | |||
AY738715.1 | - | 2, 3 | 277 | |||
AY738717.1 | - | 2, 3 | 277 | |||
AY738718.1 | - | 2, 3 | 277 | |||
AY738721.1 | - | 2, 3 | 277 | |||
S64590.1 | - | 2, 3 | 259 |