Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110850.2 | NP_001104320.1 | 3 | 248 | ||
NM_001110851.1 | NP_001104321.1 | 4 | 507 | |||
NM_001110852.1 | NP_001104322.1 | 3 | 360 | |||
NM_001110853.2 | NP_001104323.1 | 3 | 286 | |||
NM_001110856.2 | NP_001104326.1 | 4 | 395 | |||
NM_001110857.2 | NP_001104327.1 | 3 | 248 | |||
NM_001110858.2 | NP_001104328.1 | 3 | 248 | |||
NM_001110859.2 | NP_001104329.1 | 4 | 395 | |||
NM_001271503.1 | NP_001258432.1 | 4 | 395 | |||
NM_001271504.1 | NP_001258433.1 | 4 | 395 | |||
NM_001271505.1 | NP_001258434.1 | 3 | 248 | |||
NM_001271506.1 | NP_001258435.1 | 4 | 395 | |||
NM_013498.3 | NP_038526.2 | 3 | 248 | |||
GenBank mRNA | AI461618.1 | - | 4 | 357 | ||
AK009841.1 | - | 3 | 280 | |||
AK010806.1 | - | 3 | 361 | |||
AK015641.1 | - | 3 | 287 | |||
AK016156.1 | - | 3 | 273 | |||
AK076797.1 | - | 3 | 209 | |||
AK161366.1 | - | 4 | 441 | |||
AY738715.1 | - | 3 | 311 | |||
AY738716.1 | - | 2 | 164 | |||
AY738717.1 | - | 3 | 311 | |||
AY738718.1 | - | 3 | 311 | |||
AY738719.1 | - | 2 | 164 | |||
AY738720.1 | - | 4 | 407 | |||
AY738721.1 | - | 3 | 311 | |||
BC034856.1 | - | 2 | 157 | |||
M60285.1 | - | 3 | 176 | |||
S64590.1 | - | 3 | 293 |