Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110853.2 | NP_001104323.1 | 5 | 514 | ||
NM_001110859.2 | NP_001104329.1 | 6 | 623 | |||
NM_001271505.1 | NP_001258434.1 | 5 | 476 | |||
NM_001271506.1 | NP_001258435.1 | 6 | 623 | |||
GenBank mRNA | AK015641.1 | - | 5 | 515 | ||
AK016156.1 | - | 5 | 501 | |||
AK076797.1 | - | 5 | 437 | |||
AY738719.1 | - | 4 | 392 | |||
AY738721.1 | - | 5 | 539 | |||
DQ062570.1 | - | 3 | 274 |