Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018827.2 | NP_061297.1 | 6 | 1074 | ||
GenBank mRNA | AB040038.1 | - | 6 | 1074 | ||
AK160445.1 | - | 1 | 33 |