Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018827.2 | NP_061297.1 | 7 | 1279 | ||
GenBank mRNA | AB040038.1 | - | 7 | 1279 | ||
AK019030.1 | - | 2 | 272 | |||
AK160445.1 | - | 2 | 238 | |||
BC107376.1 | - | 2 | 108 | |||
BC107377.1 | - | 2 | 133 | |||
BC114407.1 | - | 2 | 103 |