Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryba4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021351.1 | NP_067326.1 | 1, 2 | 69 | ||
GenBank mRNA | AJ272228.2 | - | 1, 2 | 69 | ||
AK162018.1 | - | 2, 3 | 83 | |||
BC056444.1 | - | 1, 2 | 59 | 6824277 | ||
BC058703.1 | - | 1, 2 | 69 | 6824293 |