Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113529.1 | NP_001107001.1 | 2, 3 | 566 | ||
NM_001113530.1 | NP_001107002.1 | 2, 3 | 566 | |||
NM_007778.4 | NP_031804.3 | 2, 3 | 566 | |||
GenBank mRNA | AK136808.1 | - | 2, 3 | 340 | ||
AK138489.1 | - | 2, 3 | 340 | |||
AK154261.1 | - | 2, 3 | 567 | |||
AK154872.1 | - | 2, 3 | 340 | |||
AK160995.1 | - | 2, 3 | 340 | |||
AK166370.1 | - | 2, 3 | 339 | |||
AK170154.1 | - | 2, 3 | 276 | |||
BC025593.1 | - | 2, 3 | 352 | |||
BC066187.1 | - | 2, 3 | 320 | 30106218 | ||
BC066200.1 | - | 2, 3 | 320 | 30100524 | ||
BC066205.1 | - | 2, 3 | 320 | 30100601 | ||
CA328312.1 | - | 2, 3 | 506 | |||
M15692.1 | - | 2, 3 | 256 | |||
M21149.1 | - | 2, 3 | 229 | |||
M21952.1 | - | 2, 3 | 319 | |||
S78392.1 | - | 2, 3 | 319 | |||
X05010.1 | - | 2, 3 | 341 |