Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007780.4 | NP_031806.3 | 9 | 1361 | ||
GenBank mRNA | AF483496.1 | - | 8 | 1103 | ||
AF483497.1 | - | 8 | 1103 | |||
AK133420.1 | - | 1305 | ||||
AK152608.1 | - | 9 | 1301 | |||
AK154286.1 | - | 9 | 1298 | |||
AK155109.1 | - | 9 | 1332 | |||
AK155215.1 | - | 9 | 1241 | |||
AK155699.1 | - | 9 | 1298 | |||
AK155803.1 | - | 9 | 1298 | |||
AK155822.1 | - | 9 | 1301 | |||
AK155885.1 | - | 9 | 1302 | |||
AK171477.1 | - | 9 | 1298 | |||
BC113202.1 | - | 10 | 1372 | 40048156 | ||
M34397.1 | - | 10 | 1390 |