Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081249.1 | NP_001074718.1 | 7 | 3282 | ||
NM_001134474.1 | NP_001127946.1 | 7 | 3282 | |||
GenBank mRNA | D28599.1 | - | 7 | 3014 |