Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009977.3 | NP_034107.2 | 1 | 81 | ||
GenBank mRNA | AB015224.1 | - | 1 | 62 | ||
AK004420.1 | - | 1 | 82 | |||
AK170583.1 | - | 1 | 28 | |||
BC117047.1 | - | 1 | 66 | 40126366 | ||
BC117049.1 | - | 1 | 66 | 40126368 |