Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267695.2 | NP_001254624.2 | 8 | 1075 | ||
NM_021281.3 | NP_067256.4 | 8 | 1072 | |||
GenBank mRNA | AF038546.1 | - | 8 | 1014 | ||
AI607397.1 | - | 1 | 298 | |||
AJ223208.1 | - | 2 | 75 | |||
AK028366.1 | - | 8 | 1074 | |||
AK150274.1 | - | 8 | 1006 | |||
AK150758.1 | - | 7 | 918 | |||
AK150842.1 | - | 7 | 918 | |||
AK152191.1 | - | 8 | 1024 | |||
AK152618.1 | - | 7 | 918 | |||
AK153540.1 | - | 5 | 674 | |||
BC002125.1 | - | 8 | 981 | 3495719 | ||
BC011104.1 | - | 8 | 989 |