Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301284.1 | NP_001288213.1 | 1, 2 | 124 | ||
NM_007806.3 | NP_031832.2 | 1, 2 | 124 | |||
GenBank mRNA | AK018713.1 | - | 1, 2 | 93 | ||
AK021200.1 | - | 1, 2 | 112 | |||
AK078854.1 | - | 1, 2 | 95 | |||
AK131580.1 | - | 1, 2 | 93 | |||
AK152414.1 | - | 1, 2 | 115 | |||
AK171748.1 | - | 1, 2 | 93 | |||
BC026791.1 | - | 1, 2 | 75 | 3980538 | ||
BY005999.1 | - | 1, 2 | 127 | |||
CF738398.1 | - | 1, 2 | 107 | |||
EU791539.1 | - | 1, 2 | 54 | |||
M31775.1 | - | 1, 2 | 47 |