Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301284.1 | NP_001288213.1 | 2 | 157 | ||
NM_007806.3 | NP_031832.2 | 2 | 157 | |||
GenBank mRNA | AK018713.1 | - | 2 | 126 | ||
AK021200.1 | - | 2 | 145 | |||
AK078854.1 | - | 2 | 128 | |||
AK131580.1 | - | 2 | 126 | |||
AK152414.1 | - | 2 | 148 | |||
AK171748.1 | - | 2 | 126 | |||
BC026791.1 | - | 2 | 108 | 3980538 | ||
BC060635.1 | - | 2 | 3812 | |||
BY005999.1 | - | 2 | 160 | |||
CF738398.1 | - | 2 | 140 | |||
EU791539.1 | - | 2 | 87 | |||
M31775.1 | - | 2 | 80 |