Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276481.1 | NP_001263410.1 | 2 | 684 | ||
NM_001276482.1 | NP_001263411.1 | 3 | 597 | |||
NM_001276485.1 | NP_001263414.1 | 2 | 528 | |||
NM_001276486.1 | NP_001263415.1 | 3 | 585 | |||
NM_001276492.1 | NP_001263421.1 | 4 | 732 | |||
NM_001276493.1 | NP_001263422.1 | 3 | 689 | |||
NM_001276494.1 | NP_001263423.1 | 3 | 580 | |||
NM_010017.4 | NP_034147.1 | 3 | 741 | |||
GenBank mRNA | AK036174.1 | - | 3 | 552 | ||
AK038702.1 | - | 3 | 663 | |||
AK053886.1 | - | 2 | 489 | |||
AK145048.1 | - | 3 | 546 | |||
BC007150.1 | - | 3 | 554 | 3496914 | ||
CA327718.1 | - | 2 | 641 | |||
CD578569.1 | - | 3 | 556 | |||
X86073.1 | - | 1 | 282 |