Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037999.2 | NP_001033088.1 | 2 | 204 | ||
NM_007830.4 | NP_031856.1 | 2 | 198 | |||
GenBank mRNA | AA068307.1 | - | 2 | 144 | ||
AK008540.1 | - | 2 | 119 | |||
AK008576.1 | - | 2 | 119 | |||
AK018720.1 | - | 2 | 120 | |||
AK020692.1 | - | 2 | 168 | |||
AK027906.1 | - | 2 | 120 | |||
AK078428.1 | - | 2 | 181 | |||
AK145278.1 | - | 2 | 120 | |||
AK148753.1 | - | 2 | 120 | |||
AM000002.1 | - | 2 | 204 | |||
AV001592.1 | - | 2 | 92 | |||
BC028874.1 | - | 2 | 98 | 4194069 | ||
CJ183794.1 | - | 2 | 199 | |||
EF437365.1 | - | 2 | 105 | |||
X61431.1 | - | 2 | 117 |