Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037999.2 | NP_001033088.1 | 4 | 394 | ||
NM_007830.4 | NP_031856.1 | 4 | 388 | |||
GenBank mRNA | AA068307.1 | - | 4 | 334 | ||
AK008540.1 | - | 4 | 309 | |||
AK008576.1 | - | 4 | 309 | |||
AK018720.1 | - | 4 | 310 | |||
AK020692.1 | - | 4 | 358 | |||
AK027906.1 | - | 4 | 310 | |||
AK148753.1 | - | 4 | 310 | |||
AM000002.1 | - | 4 | 394 | |||
AV001592.1 | - | 4 | 282 | |||
BC028874.1 | - | 4 | 288 | 4194069 | ||
CJ183794.1 | - | 4 | 389 | |||
EF437365.1 | - | 4 | 295 | |||
X61431.1 | - | 4 | 307 |