Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146308.1 | NP_001139780.1 | 11 | 1086 | ||
NM_001146309.1 | NP_001139781.1 | 11 | 1074 | |||
NM_013810.3 | NP_038838.1 | 11 | 1077 | |||
GenBank mRNA | AK046073.1 | - | 11 | 1054 | ||
AK053796.1 | - | 11 | 1070 | |||
AK078082.1 | - | 11 | 1041 | |||
AK134136.1 | - | 11 | 1041 | |||
AK134487.1 | - | 11 | 1053 | |||
AK140752.1 | - | 11 | 1044 | |||
AK142818.1 | - | 11 | 1024 | |||
AK146920.1 | - | 11 | 1041 | |||
AK151096.1 | - | 11 | 1054 | |||
AK152110.1 | - | 11 | 1042 | |||
AK152204.1 | - | 11 | 1054 | |||
AK153072.1 | - | 11 | 1053 | |||
AK153389.1 | - | 11 | 1031 | |||
AK159984.1 | - | 11 | 1051 | |||
AK161973.1 | - | 10 | 1348 | |||
AK167956.1 | - | 11 | 1013 | |||
AK168898.1 | - | 11 | 1051 | |||
AK172471.1 | - | 11 | 1031 | |||
AK181313.1 | - | 2 | 154 | |||
AK210402.1 | - | 2 | 124 | |||
AY098595.1 | - | 11 | 1009 | |||
BC046430.1 | - | 11 | 1039 | 4986658 | ||
U58884.1 | - | 11 | 1030 |