Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111051.1 | NP_001104521.1 | 4 | 529 | ||
NM_001111052.1 | NP_001104522.1 | 4 | 529 | |||
NM_001195538.1 | NP_001182467.1 | 8 | 1390 | |||
NM_001195539.1 | NP_001182468.1 | 4 | 577 | |||
NM_019978.3 | NP_064362.1 | 8 | 1438 | |||
GenBank mRNA | AB093234.1 | - | 8 | 1347 | ||
AF155819.1 | - | 8 | 1336 | |||
AF155820.1 | - | 8 | 1329 | |||
AF155821.1 | - | 4 | 388 | |||
AK032424.1 | - | 4 | 419 | |||
AK047716.1 | - | 4 | 316 | |||
AK081200.1 | - | 4 | 372 | |||
AK163437.1 | - | 2 | 747 | |||
AY968047.1 | - | 4 | 277 | |||
AY968048.1 | - | 4 | 280 | |||
BC133685.1 | - | 8 | 1192 | 40110683 |