Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007861.5 | NP_031887.2 | 9 | 942 | ||
GenBank mRNA | AK117104.1 | - | 9 | 886 | ||
AK135807.1 | - | 9 | 885 | |||
AK136193.1 | - | 9 | 886 | |||
AK153399.1 | - | 9 | 886 | |||
AK167885.1 | - | 9 | 886 | |||
AK168157.1 | - | 9 | 907 | |||
AK168875.1 | - | 9 | 827 | |||
BC003368.1 | - | 9 | 869 | 3589344 | ||
U73445.1 | - | 9 | 919 |