Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010057.2 | NP_034187.1 | 3 | 850 | ||
GenBank mRNA | AB221684.1 | - | 3 | 438 | ||
AF022078.1 | - | 3 | 387 | |||
BC114342.1 | - | 3 | 434 | |||
EF535989.1 | - | 3 | 850 | |||
EF535990.1 | - | 4 | 705 | |||
U67841.1 | - | 2 | 283 |