Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110831.1 | NP_001104301.1 | 13 | 1435 | ||
NM_016878.4 | NP_058574.3 | 13 | 1259 | |||
GenBank mRNA | AK038646.1 | - | 1435 | |||
AK052001.1 | - | 1237 | ||||
AK160084.1 | - | 13 | 1196 | |||
AK166276.1 | - | 13 | 1209 | |||
AK188773.1 | - | 2 | 49 | |||
BC092232.1 | - | 13 | 1187 | 30551654 |