Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030596.3 | NP_085099.2 | 4 | 391 | ||
GenBank mRNA | AK029018.1 | - | 413 | |||
AK037142.1 | - | 4 | 422 | |||
AK148657.1 | - | 4 | 392 | |||
BC132058.1 | - | 4 | 420 | 40130335 | ||
BC132639.1 | - | 4 | 420 | 40130916 | ||
U86016.1 | - | 4 | 320 |