Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E4f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF126967.1 | - | 14 | 2286 | ||
AK033285.1 | - | 14 | 2295 | |||
AK077014.1 | - | 2398 | ||||
AK144176.1 | - | 14 | 2243 | |||
BC005634.1 | - | 14 | 2217 | |||
BC031757.1 | - | 13 | 2427 | |||
BC046459.1 | - | 13 | 2432 | 5322519 | ||
BC057011.1 | - | 13 | 1949 | |||
BC071228.1 | - | 14 | 2273 | 4915902 | ||
X76858.1 | - | 13 | 2082 |