Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007896.3 | NP_031922.1 | 4 | 591 | ||
GenBank mRNA | AK013626.1 | - | 4 | 531 | ||
AK146206.1 | - | 4 | 509 | |||
AK154245.1 | - | 4 | 535 | |||
BC043333.1 | - | 4 | 555 | |||
BC052405.1 | - | 3 | 466 | |||
BC052728.1 | - | 502 | ||||
BC064444.1 | - | 502 | 6815724 | |||
U51196.1 | - | 4 | 521 |