Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007896.3 | NP_031922.1 | 5, 6 | 738 | ||
GenBank mRNA | AK013626.1 | - | 5, 6 | 678 | ||
AK146206.1 | - | 5, 6 | 656 | |||
AK154245.1 | - | 5, 6 | 682 | |||
BC043333.1 | - | 5, 6 | 702 | |||
BC052405.1 | - | 4, 5 | 613 | |||
BC052728.1 | - | 649 | ||||
BC064444.1 | - | 649 | 6815724 | |||
U51196.1 | - | 5, 6 | 668 |