Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277925.1 | NP_001264854.1 | 2 | 900 | ||
NM_021306.3 | NP_067281.2 | 2 | 892 | |||
GenBank mRNA | AK039467.1 | - | 2 | 892 | ||
AK159365.1 | - | 2 | 902 | |||
BC057569.1 | - | 1 | 722 | 6413731 |