Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
Eef1a1
Gm6548
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
eukaryotic translation elongation factor 1 alpha 1 pseudogene
Gene Aliases:
-
EG625054
Chromosome Location:
Chr.9: 78478453 - 78481724 on Build GRCm38
Chromosome Location:
Chr.17: 78850504 - 78852544 on Build GRCm38
UniGene ID:
Mm.138471
Mm.311918
Species:
Mouse
siRNA ID
MSS274073
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Eef1a1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010106.2
NP_034236.2
6
1044
GenBank mRNA
AK032914.1
-
6
990
AK050635.1
-
6
993
AK076696.1
-
6
992
AK081725.1
-
6
992
AK083361.1
-
6
992
AK088041.1
-
6
993
AK133725.1
-
6
992
AK135448.1
-
6
993
AK137223.1
-
6
992
AK141243.1
-
6
994
AK150798.1
-
6
992
AK151479.1
-
6
993
AK151773.1
-
6
992
AK152810.1
-
6
992
AK152993.1
-
6
993
AK153387.1
-
6
992
AK153402.1
-
6
992
AK153851.1
-
6
994
AK162049.1
-
6
992
AK162313.1
-
6
991
AK162440.1
-
6
996
AK163799.1
-
6
992
AK164113.1
-
6
994
AK164671.1
-
6
992
AK165938.1
-
6
992
AK166021.1
-
6
992
AK166033.1
-
6
992
AK166058.1
-
6
992
AK166089.1
-
6
992
AK166615.1
-
6
992
AK166807.1
-
6
992
AK167843.1
-
6
992
AK167873.1
-
6
991
AK168424.1
-
6
994
AK168467.1
-
6
1000
AK190580.1
-
2
273
AK190942.1
-
2
273
AK191230.1
-
2
274
AK192548.1
-
2
273
AK192994.1
-
2
273
AK194451.1
-
2
273
AK194947.1
-
2
273
AK195943.1
-
2
273
AK197236.1
-
2
273
AK197803.1
-
2
273
AK198561.1
-
2
273
AK204290.1
-
2
291
AK212450.1
-
2
273
AK213386.1
-
2
341
AK213920.1
-
2
273
AK218081.1
-
2
273
BC003969.1
-
5
962
BC004005.1
-
5
953
3492770
BC004067.1
-
5
981
3590965
BC005660.1
-
5
982
3588966
BC018223.1
-
6
980
4012653
BC018485.1
-
6
994
3491827
BC040689.1
-
3
484
BC050124.1
-
2537
BC068258.1
-
2789
BC083069.1
-
6
980
6310448
BC092053.1
-
5
970
4008574
BC092276.1
-
5
975
3490007
BC095965.1
-
5
970
6310404
BC108391.1
-
6
977
30242277
M22432.1
-
6
983
X13661.1
-
5
960
Gene Symbol: Gm6548
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC089461.1
-
1
1291
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
translation
translational elongation
protein homooligomerization
cellular response to epidermal growth factor stimulus
tRNA binding
nucleotide binding
mRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline