Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
Eef1a1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 78478453 - 78481724 on Build GRCm38
UniGene ID:
Mm.138471
Species:
Mouse
siRNA ID
MSS274075
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Eef1a1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010106.2
NP_034236.2
4
620
GenBank mRNA
AK011771.1
-
4
568
AK012341.1
-
4
568
AK032914.1
-
4
566
AK050635.1
-
4
567
AK076696.1
-
4
568
AK081725.1
-
4
568
AK083361.1
-
4
568
AK088041.1
-
4
569
AK133725.1
-
4
568
AK135448.1
-
4
569
AK137223.1
-
4
568
AK141243.1
-
4
570
AK150798.1
-
4
568
AK151479.1
-
4
569
AK151773.1
-
4
568
AK152810.1
-
4
568
AK152993.1
-
4
569
AK153387.1
-
4
568
AK153402.1
-
4
568
AK153851.1
-
4
568
AK162049.1
-
4
568
AK162313.1
-
4
567
AK162440.1
-
4
572
AK163799.1
-
4
568
AK164113.1
-
4
570
AK164671.1
-
4
568
AK165938.1
-
4
568
AK166021.1
-
4
568
AK166033.1
-
4
568
AK166058.1
-
4
568
AK166089.1
-
4
568
AK166615.1
-
4
568
AK166807.1
-
4
568
AK167843.1
-
4
568
AK167873.1
-
4
567
AK168424.1
-
4
568
AK168467.1
-
4
574
AK187140.1
-
1
101
AK188863.1
-
2
262
AK206299.1
-
1
101
AK209036.1
-
1
99
AK210348.1
-
1
102
AK213893.1
-
1
101
BC003969.1
-
3
538
BC004005.1
-
3
529
3492770
BC004067.1
-
3
557
3590965
BC005660.1
-
3
558
3588966
BC018223.1
-
4
556
4012653
BC018485.1
-
4
570
3491827
BC040689.1
-
1
60
BC050124.1
-
2113
BC068258.1
-
2365
BC083069.1
-
4
556
6310448
BC092053.1
-
3
546
4008574
BC092276.1
-
3
551
3490007
BC092292.1
-
3
512
BC095965.1
-
3
546
6310404
BC108391.1
-
4
553
30242277
M22432.1
-
4
559
X13661.1
-
3
539
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
translation
translational elongation
protein homooligomerization
cellular response to epidermal growth factor stimulus
tRNA binding
nucleotide binding
mRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline