Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007909.3 | NP_031935.3 | 2 | 653 | ||
GenBank mRNA | AK140544.1 | - | 2 | 653 | ||
BC048697.1 | - | 2 | 393 | 6510704 | ||
U14752.1 | - | 2 | 426 | |||
U14941.1 | - | 2 | 381 |