Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010113.3 | NP_034243.2 | 2 | 571 | ||
GenBank mRNA | AK084927.1 | - | 2 | 566 | ||
AK085707.1 | - | 2 | 138 | |||
AK142666.1 | - | 2 | 267 | |||
BC017681.1 | - | 3 | 508 | 4221592 | ||
BC060741.1 | - | 2 | 556 | 4164289 | ||
BC092277.1 | - | 2 | 556 | 4240429 | ||
J00380.1 | - | 2 | 566 | |||
V00741.1 | - | 2 | 522 |