Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010119.5 | NP_034249.1 | 5 | 1385 | ||
GenBank mRNA | AF099186.1 | - | 5 | 1388 | ||
AF173156.1 | - | 5 | 1396 | |||
AK012896.1 | - | 5 | 1361 | |||
AK077536.1 | - | 5 | 1365 | |||
AK088980.1 | - | 6 | 1527 | |||
AK152821.1 | - | 5 | 1333 | |||
AK154651.1 | - | 5 | 1374 | |||
AK159191.1 | - | 5 | 1360 | |||
AK159943.1 | - | 5 | 1370 | |||
AK160535.1 | - | 5 | 1373 | |||
AK166701.1 | - | 5 | 1371 | |||
AK167721.1 | - | 5 | 1377 | |||
AK168610.1 | - | 5 | 1361 | |||
AK170679.1 | - | 5 | 1161 | |||
AK171047.1 | - | 5 | 1389 | |||
AK172539.1 | - | 5 | 1361 | |||
AK185228.1 | - | 2 | 258 | |||
AK186064.1 | - | 1 | 138 | |||
AK213323.1 | - | 1 | 148 | |||
BC018268.1 | - | 1 | 146 | |||
BC037094.1 | - | 5 | 1372 | |||
BC043332.1 | - | 5 | 1355 | 4981832 | ||
BC054828.1 | - | 5 | 1349 | 5042951 |