Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010119.5 | NP_034249.1 | 3 | 955 | ||
GenBank mRNA | AF099186.1 | - | 3 | 958 | ||
AK012896.1 | - | 3 | 931 | |||
AK077536.1 | - | 3 | 935 | |||
AK088980.1 | - | 4 | 1097 | |||
AK152821.1 | - | 3 | 904 | |||
AK154651.1 | - | 3 | 944 | |||
AK159191.1 | - | 3 | 930 | |||
AK159943.1 | - | 3 | 940 | |||
AK160535.1 | - | 3 | 943 | |||
AK166701.1 | - | 3 | 941 | |||
AK167721.1 | - | 3 | 947 | |||
AK168610.1 | - | 3 | 931 | |||
AK170679.1 | - | 3 | 731 | |||
AK171047.1 | - | 3 | 959 | |||
AK172539.1 | - | 3 | 931 | |||
AK172669.1 | - | 3 | 1117 | |||
BC037094.1 | - | 3 | 942 | |||
BC043332.1 | - | 3 | 925 | 4981832 | ||
BC054828.1 | - | 3 | 919 | 5042951 |