Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123037.2 | NP_001116509.1 | 3 | 512 | ||
NM_001123038.2 | NP_001116510.1 | 3 | 512 | |||
NM_013506.3 | NP_038534.2 | 3 | 512 | |||
GenBank mRNA | AK076509.1 | - | 3 | 223 | ||
AK088669.1 | - | 3 | 224 | |||
AK132969.1 | - | 3 | 223 | |||
AK165898.1 | - | 3 | 223 | |||
BC094422.1 | - | 3 | 208 | 6808404 | ||
CF723811.1 | - | 3 | 404 | |||
X12507.1 | - | 3 | 224 |