Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163131.1 | NP_001156603.1 | 6 | 837 | ||
NM_007921.3 | NP_031947.1 | 6 | 777 | |||
GenBank mRNA | AB221650.1 | - | 5 | 645 | ||
AF016294.1 | - | 6 | 743 | |||
AK143135.1 | - | 6 | 827 | |||
BC138692.1 | - | 6 | 741 | 8861714 | ||
BC145380.1 | - | 6 | 801 | 9053938 |