Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080388.2 | NP_001073857.1 | 11, 12 | 1748 | ||
NM_001080389.2 | NP_001073858.1 | 11, 12 | 1748 | |||
NM_001080390.2 | NP_001073859.1 | 11, 12 | 1465 | |||
NM_007928.3 | NP_031954.2 | 11, 12 | 1748 | |||
GenBank mRNA | AK045329.1 | - | 11, 12 | 1322 | ||
AK154864.1 | - | 11, 12 | 1465 | |||
AK172444.1 | - | 11, 12 | 1708 | |||
AK172664.1 | - | 11, 12 | 1391 | |||
AK211690.1 | - | 2, 3 | 196 | |||
AK220191.1 | - | 12, 13 | 1314 | |||
BC058556.1 | - | 11, 12 | 1685 | 5289799 | ||
CX210531.1 | - | 1, 2 | 111 |