Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290753.1 | NP_001277682.1 | 14, 15 | 2884 | ||
NM_010142.3 | NP_034272.1 | 14, 15 | 2881 | |||
GenBank mRNA | BC043088.1 | - | 14, 15 | 2805 | 6418088 | |
BC062924.1 | - | 14, 15 | 2829 | |||
L25890.1 | - | 15, 16 | 2723 | |||
X76011.1 | - | 8, 9 | 1182 |