Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042630.2 | NP_001036095.1 | 4 | 451 | ||
NM_001276257.1 | NP_001263186.1 | 4 | 451 | |||
NM_007947.3 | NP_031973.2 | 4 | 451 | |||
GenBank mRNA | AK021014.1 | - | 4 | 449 | ||
AK136629.1 | - | 4 | 454 | |||
AK162355.1 | - | 4 | 450 | |||
U68546.1 | - | 4 | 347 |