Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253392.1 | NP_001240321.1 | 21 | 2820 | ||
NM_007958.1 | NP_031984.1 | 23 | 3281 | |||
GenBank mRNA | AK122454.1 | - | 23 | 3088 | ||
BC042442.1 | - | 21 | 2813 | 3980726 | ||
CF531941.1 | - | 5 | 564 | |||
X69942.1 | - | 23 | 3281 |